COMH93D64DA2 Combi-Blocks, Inc.

1-Bromo-3-(difluoromethyl)-4-fluoro-2-methoxy-benzene

  • MDL number MFCD29767789
  • Cas Number : 2091158-36-8
  • Molecular Weight : 255.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-8740

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-8740
Availability details for
Quantity
  • Availability
  • Add to Cart