COMH93D6505E Combi-Blocks, Inc.

Mal-peg3-alcohol

  • MDL number MFCD28142465
  • Cas Number : 146551-23-7
  • Molecular Weight : 229.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-9557

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-9557
Availability details for
Quantity
  • Availability
  • Add to Cart