COMH93D6505F Combi-Blocks, Inc.

7-((Benzyloxy)methyl)-1,4-oxazepane

  • MDL number MFCD30002165
  • Cas Number : 1926190-22-8
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-9558

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-9558
Availability details for
Quantity
  • Availability
  • Add to Cart