COMH93D650C1 Combi-Blocks, Inc.

N-(3,5-Difluorobenzyl)-n-[(1,3-dimethyl-1h-pyrazol-4-yl)methyl]amine

  • MDL number MFCD08696806
  • Cas Number : 1006436-76-5
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-9665

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-9665
Availability details for
Quantity
  • Availability
  • Add to Cart