COMH93D650CB Combi-Blocks, Inc.

3-Fluoropyridine-2-carbonyl chloride

  • MDL number MFCD25969154
  • Cas Number : 929568-98-9
  • Molecular Weight : 159.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-9677

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-9677
Availability details for
Quantity
  • Availability
  • Add to Cart