COMH93D651FF Combi-Blocks, Inc.

1-(5-Bromonaphthalen-2-yl)cyclopropane-1-carbonitrile

  • MDL number MFCD30496597
  • Cas Number : 2116763-10-9
  • Molecular Weight : 272.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HG-0746

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HG-0746
Availability details for
Quantity
  • Availability
  • Add to Cart