COMH93D6524A Combi-Blocks, Inc.

[8]Cycloparaphenylene

  • MDL number MFCD32062876
  • Cas Number : 1217269-85-6
  • Molecular Weight : 608.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HG-2253

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HG-2253
Availability details for
Quantity
  • Availability
  • Add to Cart