COMH93D65417 Combi-Blocks, Inc.

1-(Tetrahydrofuran-2-ylmethyl)piperidin-4-amine

  • MDL number MFCD09047072
  • Cas Number : 108282-26-4
  • Molecular Weight : 184.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?JA-3542

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?JA-3542
Availability details for
Quantity
  • Availability
  • Add to Cart