COMH93D65AEC Combi-Blocks, Inc.

2-Bromo-1-(3,4-dihydro-2h-1,5-benzodioxepin-7-yl)ethan-1-one

  • MDL number MFCD00218462
  • Cas Number : 35970-34-4
  • Molecular Weight : 271.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4130

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4130
Availability details for
Quantity
  • Availability
  • Add to Cart