COMH93D65B2A Combi-Blocks, Inc.

(S)-N4-Benzyl-2-isobutylpiperazine

  • MDL number MFCD03787915
  • Cas Number : 444892-03-9
  • Molecular Weight : 232.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5920

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5920
Availability details for
Quantity
  • Availability
  • Add to Cart