COMH93D65B34 Combi-Blocks, Inc.

4,4'-Azoxydiphenetole

  • MDL number MFCD00026790
  • Cas Number : 4792-83-0
  • Molecular Weight : 286.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6208

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6208
Availability details for
Quantity
  • Availability
  • Add to Cart