COMH93D65B3C Combi-Blocks, Inc.

2-Chloro-4,5-difluorophenylacetic acid

  • MDL number MFCD04973421
  • Cas Number : 883502-00-9
  • Molecular Weight : 206.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6413

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6413
Availability details for
Quantity
  • Availability
  • Add to Cart