COMH93D65B4C Combi-Blocks, Inc.

2,3-Dihydro-1h-pyrrolo[3,4-c]pyridine

  • MDL number MFCD06858479
  • Cas Number : 496-13-9
  • Molecular Weight : 120.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7040

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7040
Availability details for
Quantity
  • Availability
  • Add to Cart