COMH93D65B5A Combi-Blocks, Inc.

2-Mercapto-5-n-propylpyrimidine

  • MDL number MFCD07777095
  • Cas Number : 52767-84-7
  • Molecular Weight : 154.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7353

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7353
Availability details for
Quantity
  • Availability
  • Add to Cart