COMH93D65B5E Combi-Blocks, Inc.

4-(1,3-Thiazol-2-yl)benzoic acid

  • MDL number MFCD08275675
  • Cas Number : 266369-49-7
  • Molecular Weight : 205.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7681

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7681
Availability details for
Quantity
  • Availability
  • Add to Cart