COMH93D65B76 Combi-Blocks, Inc.

3-Bromo-5-fluorobenzylamine, HCl

  • MDL number MFCD04113608
  • Cas Number : 1189924-80-8
  • Molecular Weight : 240.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2167

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2167
Availability details for
Quantity
  • Availability
  • Add to Cart