COMH93D65C24 Combi-Blocks, Inc.

5-Fluoro-6-methoxypyridine-3-boronic acid pinacol ester

  • MDL number MFCD07374975
  • Molecular Weight : 253.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8460

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-8460
Availability details for
Quantity
  • Availability
  • Add to Cart