COMH93D65CC9 Combi-Blocks, Inc.

Dirithromycin

  • MDL number MFCD00865041
  • Cas Number : 62013-04-1
  • Molecular Weight : 835.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1R,2R,3R,6R,7S,8S,9R,10R,12R,13S,15R,17S)-3-ethyl-2,10-dihydroxy-15-[(2-methoxyethoxy)methyl]-2,6,8,10,12,17-hexamethyl-5-oxo-9-{[3,4,6-trideoxy-3-(dimethylamino)-beta-D-xylo-hexopyranosyl]oxy}-4,16-dioxa-14-azabicyclo[11.3.1]heptadec-7-yl 2,6-dideoxy-3-
Inchi Code 1S/C42H78N2O14/c1-15-29-42(10,49)37-24(4)32(43-30(56-37)21-52-17-16-50-13)22(2)19-40(8,48)36(58-39-33(45)28(44(11)12)18-23(3)53-39)25(5)34(26(6)38(47)55-29)57-31-20-41(9,51-14)35(46)27(7)54-31/h22-37,39,43,45-46,48-49H,15-21H2,1-14H3/t22-,23-,24+,25+,26-,27+,28+,29-,30-,31+,32+,33-,34+,35+,36-,37-,39+,40-,41-,42-/m1/s1
Inchi Key WLOHNSSYAXHWNR-DWIOZXRMSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7657

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7657
Availability details for
Quantity
  • Availability
  • Add to Cart