COMH93D65CF3 Combi-Blocks, Inc.

6-T-Butylnaphthalen-2-boronic acid pinacol ester

  • MDL number MFCD20527113
  • Cas Number : 1620789-31-2
  • Molecular Weight : 310.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-2918

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-2918
Availability details for
Quantity
  • Availability
  • Add to Cart