COMH93D65D35 Combi-Blocks, Inc.

Mc-mmaf

  • MDL number MFCD25976741
  • Cas Number : 863971-19-1
  • Molecular Weight : 925.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9735

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9735
Availability details for
Quantity
  • Availability
  • Add to Cart