COMH93D65D3D Combi-Blocks, Inc.

2,4,6-Trimethylphenyl isothiocyanate

  • MDL number MFCD00022055
  • Cas Number : 6095-82-5
  • Molecular Weight : 177.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0024

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0024
Availability details for
Quantity
  • Availability
  • Add to Cart