COMH93D65D6A Combi-Blocks, Inc.

4'-Ethoxy-2',3'-difluoroacetophenone

  • MDL number MFCD09258710
  • Cas Number : 1017779-68-8
  • Molecular Weight : 200.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0195

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0195
Availability details for
Quantity
  • Availability
  • Add to Cart