COMH93D65D98 Combi-Blocks, Inc.

Diethoxymethylvinylsilane

  • MDL number MFCD00009067
  • Cas Number : 5507-44-8
  • Molecular Weight : 160.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name diethoxy(methyl)vinylsilane
Inchi Code 1S/C7H16O2Si/c1-5-8-10(4,7-3)9-6-2/h7H,3,5-6H2,1-2,4H3
Inchi Key MBGQQKKTDDNCSG-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0374

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0374
Availability details for
Quantity
  • Availability
  • Add to Cart