COMH93D65D9B Combi-Blocks, Inc.

4-Ethoxy-3-(trifluoromethyl)benzyl alcohol

  • MDL number MFCD13196581
  • Cas Number : 1206593-33-0
  • Molecular Weight : 220.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0380

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0380
Availability details for
Quantity
  • Availability
  • Add to Cart