COMH93D6623A Combi-Blocks, Inc.

Dimethylethoxysilane

  • MDL number MFCD00026750
  • Cas Number : 14857-34-2
  • Molecular Weight : 104.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0705

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-0705
Availability details for
Quantity
  • Availability
  • Add to Cart