COMH93D6630F Combi-Blocks, Inc.

1,2-Phenylenedioxydiacetic acid

  • MDL number MFCD00014353
  • Cas Number : 5411-14-3
  • Molecular Weight : 226.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [2-(carboxymethoxy)phenoxy]acetic acid
Inchi Code 1S/C10H10O6/c11-9(12)5-15-7-3-1-2-4-8(7)16-6-10(13)14/h1-4H,5-6H2,(H,11,12)(H,13,14)
Inchi Key PPZYHOQWRAUWAY-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-1452

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-1452
Availability details for
Quantity
  • Availability
  • Add to Cart