COMH93D6639F Combi-Blocks, Inc.

Dimethyl (phthalimidomethyl)phosphonate

  • MDL number MFCD00015464
  • Cas Number : 28447-26-9
  • Molecular Weight : 269.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-1952

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-1952
Availability details for
Quantity
  • Availability
  • Add to Cart