COMH93D663BF Combi-Blocks, Inc.

(1R,2R)-(-)-2-Benzyloxycyclohexyl isothiocyanate

  • MDL number MFCD05664116
  • Cas Number : 745783-98-6
  • Molecular Weight : 247.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2056

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2056
Availability details for
Quantity
  • Availability
  • Add to Cart