COMH93D663C6 Combi-Blocks, Inc.

1-Octylphosphonic dichloride

  • MDL number MFCD00015813
  • Cas Number : 3095-94-1
  • Molecular Weight : 231.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2074

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2074
Availability details for
Quantity
  • Availability
  • Add to Cart