COMH93D663F3 Combi-Blocks, Inc.

4-Hydroxyphenyl benzoate

  • MDL number MFCD00053304
  • Cas Number : 2444-19-1
  • Molecular Weight : 214.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2241

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2241
Availability details for
Quantity
  • Availability
  • Add to Cart