COMH93D663FA Combi-Blocks, Inc.

3-(2-Pyridyl)-5,6-bis(4-sulfophenyl)-1,2,4-triazine disodium salt hydrate

  • MDL number MFCD00717621
  • Cas Number : 28048-33-1
  • Molecular Weight : 532.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2260

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2260
Availability details for
Quantity
  • Availability
  • Add to Cart