COMH93D66419 Combi-Blocks, Inc.

Diethyl 1-tetradecanephosphonate

  • MDL number MFCD00015597
  • Cas Number : 5191-09-3
  • Molecular Weight : 334.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2349

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2349
Availability details for
Quantity
  • Availability
  • Add to Cart