COMH93D66430 Combi-Blocks, Inc.

2,5-Dibromohexane

  • MDL number MFCD00039177
  • Cas Number : 24774-58-1
  • Molecular Weight : 243.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2424

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2424
Availability details for
Quantity
  • Availability
  • Add to Cart