COMH93D664A5 Combi-Blocks, Inc.

4-N-Nonyloxybenzaldehyde

  • MDL number MFCD00043712
  • Cas Number : 50262-46-9
  • Molecular Weight : 248.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2818

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2818
Availability details for
Quantity
  • Availability
  • Add to Cart