COMH93D664BA Combi-Blocks, Inc.

Diethyl dipropylmalonate

  • MDL number MFCD00026840
  • Cas Number : 6065-63-0
  • Molecular Weight : 244.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2887

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2887
Availability details for
Quantity
  • Availability
  • Add to Cart