COMH93D664BE Combi-Blocks, Inc.

Isopropyl trifluoroacetate

  • MDL number MFCD00013566
  • Cas Number : 400-38-4
  • Molecular Weight : 156.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name isopropyl trifluoroacetate
Inchi Code 1S/C5H7F3O2/c1-3(2)10-4(9)5(6,7)8/h3H,1-2H3
Inchi Key ASAXRKSDVDALDT-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2901

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2901
Availability details for
Quantity
  • Availability
  • Add to Cart