COMH93D664C7 Combi-Blocks, Inc.

Diisoamylamine

  • MDL number MFCD00015053
  • Cas Number : 544-00-3
  • Molecular Weight : 157.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2935

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2935
Availability details for
Quantity
  • Availability
  • Add to Cart