COMH93D664D3 Combi-Blocks, Inc.

2-Valerylfuran

  • MDL number MFCD00053147
  • Cas Number : 3194-17-0
  • Molecular Weight : 152.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2979

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2979
Availability details for
Quantity
  • Availability
  • Add to Cart