COMH93D664F8 Combi-Blocks, Inc.

1-(2,5-Dimethoxyphenyl)-2-thiourea

  • MDL number MFCD00022163
  • Cas Number : 67617-98-5
  • Molecular Weight : 212.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(2,5-dimethoxyphenyl)thiourea
Inchi Code 1S/C9H12N2O2S/c1-12-6-3-4-8(13-2)7(5-6)11-9(10)14/h3-5H,1-2H3,(H3,10,11,14)
Inchi Key ZLGQKHQZDDTIFW-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3089

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3089
Availability details for
Quantity
  • Availability
  • Add to Cart