COMH93D66515 Combi-Blocks, Inc.

Methyl 4-n-butoxybenzoate

  • MDL number MFCD00070801
  • Cas Number : 4906-25-6
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-butoxybenzoate
Inchi Code 1S/C12H16O3/c1-3-4-9-15-11-7-5-10(6-8-11)12(13)14-2/h5-8H,3-4,9H2,1-2H3
Inchi Key PUHCYTHGLBCWEA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3177

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3177
Availability details for
Quantity
  • Availability
  • Add to Cart