COMH93D66533 Combi-Blocks, Inc.

3,5-Dimethyl-3-heptanol

  • MDL number MFCD00046585
  • Cas Number : 19549-74-7
  • Molecular Weight : 144.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,5-dimethyl-3-heptanol
Inchi Code 1S/C9H20O/c1-5-8(3)7-9(4,10)6-2/h8,10H,5-7H2,1-4H3
Inchi Key NOSOEGQGQOMHBF-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3283

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3283
Availability details for
Quantity
  • Availability
  • Add to Cart