COMH93D6653C Combi-Blocks, Inc.

1-Ethyl-3-methylpiperidin-4-one

  • MDL number MFCD01318144
  • Cas Number : 3612-16-6
  • Molecular Weight : 141.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-ethyl-3-methyl-4-piperidinone
Inchi Code 1S/C8H15NO/c1-3-9-5-4-8(10)7(2)6-9/h7H,3-6H2,1-2H3
Inchi Key DZPMPRBBBQMILW-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3314

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3314
Availability details for
Quantity
  • Availability
  • Add to Cart