COMH93D6653D Combi-Blocks, Inc.

Tetraheptylammonium iodide

  • MDL number MFCD00041984
  • Cas Number : 3535-83-9
  • Molecular Weight : 537.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N,N,N-triheptyl-1-heptanaminium iodide
Inchi Code 1S/C28H60N.HI/c1-5-9-13-17-21-25-29(26-22-18-14-10-6-2,27-23-19-15-11-7-3)28-24-20-16-12-8-4;/h5-28H2,1-4H3;1H/q+1;/p-1
Inchi Key KCSOHLKZTZMKQA-UHFFFAOYSA-M

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3324

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3324
Availability details for
Quantity
  • Availability
  • Add to Cart