COMH93D6654A Combi-Blocks, Inc.

3,4-Dimethyl-1-pentyn-3-ol

  • MDL number MFCD00039845
  • Cas Number : 1482-15-1
  • Molecular Weight : 112.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,4-dimethyl-1-pentyn-3-ol
Inchi Code 1S/C7H12O/c1-5-7(4,8)6(2)3/h1,6,8H,2-4H3
Inchi Key DZNLEQBXXLGELU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3370

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3370
Availability details for
Quantity
  • Availability
  • Add to Cart