COMH93D66553 Combi-Blocks, Inc.

Hexyl methyl ether

  • MDL number MFCD00015350
  • Cas Number : 4747-07-3
  • Molecular Weight : 116.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name hexyl methyl ether
Inchi Code 1S/C7H16O/c1-3-4-5-6-7-8-2/h3-7H2,1-2H3
Inchi Key ICBJCVRQDSQPGI-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3398

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3398
Availability details for
Quantity
  • Availability
  • Add to Cart