COMH93D66563 Combi-Blocks, Inc.

1H-Pyrazole-1-(n-methylcarboxamidine) hydrochloride

  • MDL number MFCD11114408
  • Cas Number : 194852-88-5
  • Molecular Weight : 160.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-methyl-1H-pyrazole-1-carboximidamide hydrochloride
Inchi Code 1S/C5H8N4.ClH/c1-7-5(6)9-4-2-3-8-9;/h2-4H,1H3,(H2,6,7);1H
Inchi Key SYSWHVMZAHBYJO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3479

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3479
Availability details for
Quantity
  • Availability
  • Add to Cart