COMH93D66574 Combi-Blocks, Inc.

4,4'-Dichlorochalcone

  • MDL number MFCD00018704
  • Cas Number : 19672-59-4
  • Molecular Weight : 277.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3524

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3524
Availability details for
Quantity
  • Availability
  • Add to Cart