COMH93D66579 Combi-Blocks, Inc.

Pinacolone oxime

  • MDL number MFCD00019956
  • Cas Number : 2475-93-6
  • Molecular Weight : 115.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2E)-3,3-dimethyl-2-butanone oxime
Inchi Code 1S/C6H13NO/c1-5(7-8)6(2,3)4/h5H,1-4H3
Inchi Key YVEBKIYPCFXHDE-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3542

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3542
Availability details for
Quantity
  • Availability
  • Add to Cart