COMH93D66594 Combi-Blocks, Inc.

1-Cyclohexyl-1-butanol

  • MDL number MFCD00019360
  • Cas Number : 4352-42-5
  • Molecular Weight : 156.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3648

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3648
Availability details for
Quantity
  • Availability
  • Add to Cart