COMH93D665AE Combi-Blocks, Inc.

Stearolic acid

  • MDL number MFCD00014386
  • Cas Number : 506-24-1
  • Molecular Weight : 280.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 9-octadecynoic acid
Inchi Code 1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-8,11-17H2,1H3,(H,19,20)
Inchi Key RGTIBVZDHOMOKC-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3758

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3758
Availability details for
Quantity
  • Availability
  • Add to Cart